Learn More
Cole-Parmer Digi-Sense J20-5-502 Thermocouple, Wire, Type-J, 20-Gauge, Polyvinyl Insulation, 1000-ft Spool
Supplier:  Cole-Parmer 0854215
Encompass_Preferred
Wire comes in 20-gauge size, ideal for fabricating your own probes or extension cables (meets ANSI and ASTM standards). Choose from wire with polyvinyl chloride (PVC), fluorinated ethylene propylene resin (FEP/FEP), fiberglass, high-temperature fiberglass, or ceramic fiber outer/inner insulation. These cables are also available in 100-ft or 1000-ft lengths for any application.
Catalog No. 50-196-1535