Learn More
Cole-Parmer Argos Technologies Syringe Filter, 30mm, .22um, Red PP/Surfactant-Free Cellulose Acetate; 50/Pk
Supplier:  Cole-Parmer 0439592
Encompass
Choose the membrane material best suited for your application. Each filter is quality tested for filter efficiency and housing integrity for use up to 75 psi (5.1 bar). Sterile filters are certified nonpyrogenic, have been gamma sterilized, and come individually packaged with lot numbers printed on for easy traceability.
Catalog No. 50-196-1400
Due to product restrictions, please to purchase or view availability for this product.