Learn More
Synthware™ Filter Funnels
Used in conjunction with vacuum source to filter and collect fluids
Supplier: Synthware™ F752490M
Description
- With standard taper inner joint and hose connector for connection to vacuum source
- Hose connection O.D.: 24 mm
Specifications
Porosity: Medium | |
40 to 60 μm | |
90 mm |
Borosilicate Glass (3.3) | |
Filter Funnel |