Learn More
Syngene™ UV Transilluminator 2530LM
SDP
Syngene™ UV Transilluminator 2530LM for illumination of any sample excited by UV light.
Supplier:  Syngene™ GX2530LME
Catalog No. 01-257-270
Add to Cart
Description
Description
  • Sealed to allow wet gels on surface
  • For use in Syngene™ Systems
  • 365/302nm wavelength
Specifications
Specifications
365/302 nm
UV Transilluminator
230 w
335 x 310 x 100 mm
250 x 300 mm
230 V
Fixed
For G:Box systems
250 x 300 mm
230 V 50 Hz 230 w
Syngene 2530LM
50 Hz