Learn More
Fisher Science Education™ Motic D-Moticam 1080 Digital HDMI Microscopy Camera
To capture high resolution video and images directly onto an SD card (not included) at up to 2.0 MP.
Description
- Multi-tasking camera for educational, industrial or clinical use
- For use with HDMI direct output or capture to SD card (not included)
- Interface connection: HDMI (1080) AV and USB
- Two adapters to fit microscope eyepiece O.D. 30mm and 38mm
Specifications
Microscope Camera | |
1280x960 (1.3MP) at15 fps under optimal illumination conditions | |
CMOS |