Learn More
Med Vet International Ethicon Vicryl Rapide, 4-0 Polyglactin Suture, 18in. P-3, 12/box
Supplier: Med Vet International VR494
The Ethicon VR494 suture is a high-quality absorbable Polyglactin 910 surgical suture. It measures 18in. in length, has a size of 4-0, they are dyed black to provide greater visibility while suturing, and typically have an absorption rate of 56-70 days. This suture comes with a P-3 Precision Point reverse cutting needle and has 12 sutures in a box.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.