Learn More
Surgical Design No.25 Stainless Steel Scalpel Blade
Straight edged blade fits handles 4, 4L, 4 Graduated and 6B
Supplier: Surgical Design BLADE#25S
Description
- More sharply angled back edge making it almost equilateral in shape
- For precision cutting in micro-circuitry, masks and friskets
- Similar but much larger than a no.11 blade
Specifications
# 25 | |
Handles # 4, # 4L, # 4 Graduated and # 6B |