Learn More
Surgical Design No. 23 Carbon Scalpel Blade
Leaf-shaped blade fits handles 4, 4L, 4 Graduated and 6B
Supplier: Surgical Design BLADE#23
Description
Surgical Design™ presents Royaltek™- our premium line of surgical blades- Exceptionally strong and unifoirmly sharpened to maximum acuity
- Surgical grade, carbon-steel blades
- Individually sealed
- Peelable packing
- Sterile
Specifications
# 23 | |
Handles # 4, # 4L, # 4 Graduated and # 6B |