Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher Healthcare™ Sure-Vue™ RPR Test Kit
Specifications
Detectable Analytes | Reagin (Syphilis) |
---|---|
For Use With (Application) | For in-vitro diagnostic use |
Includes | Reagent, prediluted positive and negative control, dispensing pipets, needle, Plastic dispensing vial and disposable slides |
Includes: Reagent, prediluted positive and negative controls, dispensing pipets, needle, plastic dispensing vial, disposable slides, and package insert.
Catalog Number | Quantity | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Quantity | Description | Price | Quantity & Availability | |||||
23-038020
|
5000 Tests | RPR Test Kit, 30-well |
Pack of 5000 for $7,723.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
23-038021
|
5000 Tests | RPR Test Kit, 10-well |
Pack of 5000 for $3,140.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
23-038009
|
100 Tests | RPR Test Kit |
Pack of 100 for $184.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
23-038010
|
500 Tests | RPR Test Kit |
Pack of 500 for $539.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
- Antigen detects reagin present in serum of syphilitic patients
- For in vitro diagnostic use
Specifications
Reagin (Syphilis) | |
For in-vitro diagnostic use | |
Reagent, prediluted positive and negative control, dispensing pipets, needle, Plastic dispensing vial and disposable slides |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title