Learn More
Andwin Scientific SUPOR 450 PES .45U 47MM 100/PK
Supplier: Andwin Scientific 28147640
The Supor Membrane Disc Filter has a pore size of 0.45 m and a diameter of 47 mm. This product has a hydrophilic polyethersulfone membrane that features high flow rate, low protein binding, and low extractables. It is stronger than other types of membranes, namely acetate and nitrate. This product also features a very high water flow fluid filtration capacity. This is sold in a pack of 100.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.