Learn More
Andwin Scientific SUPOR 200 0.2UM 47MM 100PK
Supplier: Andwin Scientific 60301
The Supor 200 PES Membrane Disc Filters are ideal for use in biological, pharmaceutical, and sterilizing filtration applications. This product provides quick filtration with the best flow rates and high throughputs. The filters feature low protein binding and compatibility with a wide range of drugs for demanding procedures. This product has a diameter of 47 mm and a pore size of 0.2 m, and is sold in a pack of 100.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.