Learn More
MilliporeSigma™ Supelco™ Supelpure™ HC Hydrocarbon Trap, S-Trap
Activated charcoal adsorbs hydrocarbons and other contaminants from carrier gases, air, and hydrogen
Supplier:  MilliporeSigma™ Supelco™ 503142
Catalog No. 11-100-9702
Add to cart
Description
Description
  • Operates efficiently for approximately six months when total hydrocarbons in the incoming gas average 10 ppm
Specifications
Specifications
S-Trap
1/8 in.
19 cm
14.1 cm
13 mm
Hydrocarbon/Contaminant Removal
7.5 in.
5.6 in.
0.5 in.
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Supelpure is a registered trademark of Sigma-Aldrich Co. LLC