Learn More
MilliporeSigma™ Supelco™ SupelMIP™ SPE Tube - Full Beta-receptor (beta-blockers and beta-agonists), Bed Wt. 25 mg
Solid phase extraction (SPE) is a technique commonly used in laboratories to isolate analytes of interest from complex matrices.
Manufacturer:  MilliporeSigma™ Supelco™ 53224U
Catalog No. 11-100-7268
Add to cart
Description
Description
SupelMIP SPE phases were developed by MIP Technologies AB, which is one of the leading authorities and commercial pioneers of molecular imprinted polymers for process scale separations, analytical chromatography, and sample preparation. The SupelMIP SPE line consist s of highly cross-linked polymers that are engineered to extract a single analyte of interest or a class of structurally related analytes with an extremely high degree of selectivity. This is possible because selectivity is introduced during MIP synthesis in which a template molecule, designed to mimic the analyte, guides the formation of specific cavities or imprints that are sterically and chemically complementary to the analyte(s) of interest.
By careful design of the imprinting site, either by molecular modeling, experimental design, or screening methods, the binding cavities can be engineered to offer multiple interaction points with the analyte(s) of interest. This leads to a stronger interaction between the solid-phase and the analyte(s). As a consequence, harsher wash conditions can be tolerated during SPE methodology resulting in cleaner extracts. Because extraction selectivity is significantly improved, lower background is observed allowing analysts to achieve lower limits of detection. 
  • Highly selective SPE for trace analysis in complex matrices.
  • Achieve lower detection limits.
  • Significant time and cost savings.
  • Improved MS-compatibility
Specifications
Specifications
Full Beta-receptor (beta-blockers and beta-agonists)
25 mg
SPE Column
Polypropylene
SPE
SupelMIP
3 mL
50/Pk.
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
SupelMIP is a registered trademark of Sigma-Aldrich Co. LLC