Learn More
MilliporeSigma™ Supelco™ SupelMIP™ SPE Tube - Beta-agonists, Bed Wt. 25 mg
Solid phase extraction (SPE) is a technique commonly used in laboratories to isolate analytes of interest from complex matrices.
$729.66
Specifications
Bed Weight 25 mg
Column Type SPE
Product Line SupelMIP
Column Phase Beta-agonists
Material Polypropylene
View More Specs
Products 2
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
11-100-8089
View Documents
MilliporeSigma™ Supelco™
53225U
3 mL
Each for $729.66
Add to cart
11-100-8088
View Documents
MilliporeSigma™ Supelco™
53202U
10 mL
Each for $729.66
Add to cart
Description
Description
SupelMIP SPE phases were developed by MIP Technologies AB, which is one of the leading authorities and commercial pioneers of molecular imprinted polymers for process scale separations, analytical chromatography, and sample preparation. The SupelMIP SPE line consist s of highly cross-linked polymers that are engineered to extract a single analyte of interest or a class of structurally related analytes with an extremely high degree of selectivity. This is possible because selectivity is introduced during MIP synthesis in which a template molecule, designed to mimic the analyte, guides the formation of specific cavities or imprints that are sterically and chemically complementary to the analyte(s) of interest.
By careful design of the imprinting site, either by molecular modeling, experimental design, or screening methods, the binding cavities can be engineered to offer multiple interaction points with the analyte(s) of interest. This leads to a stronger interaction between the solid-phase and the analyte(s). As a consequence, harsher wash conditions can be tolerated during SPE methodology resulting in cleaner extracts. Because extraction selectivity is significantly improved, lower background is observed allowing analysts to achieve lower limits of detection. 
  • Highly selective SPE for trace analysis in complex matrices.
  • Achieve lower detection limits.
  • Significant time and cost savings.
  • Improved MS-compatibility
Specifications
Specifications
25 mg
SupelMIP
Polypropylene
SPE
Beta-agonists
SPE Column
SDS
Product Certifications
SupelMIP is a registered trademark of Sigma-Aldrich Co. LLC