Learn More
MilliporeSigma™ Supelco™ Supel™Genie RP-Amide Online SPE Cartridges, Bed Wt. 180 mg
Solid phase extraction (SPE) is a technique commonly used in laboratories to isolate analytes of interest from complex matrices.
$967.29 - $2322.01
Specifications
Bed Weight 180 mg
Column Phase RP-Amide
Product Line Supel
Column Type SPE
For Use With (Application) Food and Beverages
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-100-7193
View Documents
MilliporeSigma™ Supelco™
55519U
2/Pk.
Each for $967.29
Add to cart
11-100-5983
View Documents
MilliporeSigma™ Supelco™
55522U
6/Pk.
Each for $2,322.01
Add to cart
Description
Description
  • Provides cleaner samples for longer column life, less instrument downtime, and more accurate and reproducible data.
  • Automation saves time, decreases costs, and eliminates human error.
  • Highly reproducible, consistent product performance.
  • RP-Amide cartridges often offer improved performance for polar analytes, especially those that can interact via hydrogen bonding, when compared to standard alkyl phases.
  • Cartridges also available in pack of 6
Specifications
Specifications
180 mg
Supel
Food and Beverages
SPE Column
RP-Amide
SPE
Stainless steel
SDS
Product Certifications
Supel is a trademark of Sigma-Aldrich Co. LLC