Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sulfacetamide Sodium, Monohydrate, USP, 99-100.5%, Spectrum™ Chemical

Click to view available options
Quantity:
100 g
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Description
Spectrum™ Chemical Sulfacetamide Sodium, Monohydrate, USP is an antibiotic that is used to treat skin infections and urinary tract. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
CAS Max % | 1 |
pH | 8.0 to 9.5 |
Packaging | Fiber Drum |
Quantity | 50 kg |
Percent Purity | 99 to 100.5% |
Grade | USP |
Water | 0.081 |