Learn More
Sigma Aldrich Fine Chemicals Biosciences Suba-Seal(R) septa red rubber, Suba Seal, 13, fits neck I.D., 9.5 mm, for use with 10/30 joints | 100EA
Supplier: Sigma Aldrich Fine Chemicals Biosciences Z124567100EA
Suba-Seal(R) septa red rubber, Suba Seal, 13, fits neck I.D., 9.5 mm, for use with 10/30 joints | 100EA