Learn More
Carolina™ Student Plant Press
For use in drying and flattening plant samples. Carolina™ Student Plant Press is used in study of preserved botany.
Supplier: Carolina™ 663060
Alert:
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Description
Includes:- Two press boards (12 × 16 in.) constructed of latticed pine strips with 5/8 in. double-prong staples at all joints
- Two 3/4 × 52 in. straps
- 12 botanical corrugated ventilators (12 x 16 in.)
- 12 blotting-paper drier
Specifications
Plant Press | |
2 press boards, 2 straps, 12 corrugated ventilators, 12 blotting-paper driers | |
12 x 16 in. | |
5 lb. |