Learn More
Cole-Parmer Stuart SMP10 Digital Melting Point Apparatus, 120 VAC
Supplier:  Cole-Parmer 0301140
Encompass_Preferred
This melting point apparatus lets you select, measure and display the temperature digitally, ensuring accuracy and repeatability. Easily view the melt using the illuminated magnifying lens. The apparatus allows you to be more productive by testing multiple samples at the same time. Save time, too, by setting a plateau temperature for its initial warm-up. The model, SMP10 provides 300C maximum temperature with 1resolution. It operates at 120 VAC.
Catalog No. NC1113333