Learn More
Streck UA-Cellular™ Complete Combined Cellular and Chemistry Urinalysis Control
Urinalysis control designed for Siemens CLINITEK Atlas and Sysmex UF-1000i combined urinalysis system
Supplier: Streck 218991
This item is not returnable.
View return policy
Description
- Walk-away capability
- Tests entire Siemens CLINITEK AUWi system's ability to accurately identify and quantify chemistry and cellular components
- Designed to align six stabilized urine particle types (RBC, WBC, cast, crystals, bacteria and epithelial) with common urine chemistry analytes found in patient sample
- No setting changes required to move between patient and control modes
- Stability:
- 105-day closed-vial stability
- 30-day open-vial stability
Specifications
Siemens CLINITEK Atlas and Sysmex UF-1000i combined urinalysis system | |
3 x 120 mL (L1-PC, L1-PCM, L-NC) |