Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Streck Cal-Chex A Plus Calibrator
Hematology calibrator assayed for Abbott CELL-DYN™ 3200, 3500 and 3700 analyzers
Specifications
For Use With (Equipment) | Abbott CELL-DYN™ 1700, 3000, 3200, 3500 and 3700 analyzer |
---|---|
Type | Cal-Chex A Plus Hematology Calibrators |
Stability | 5 Days Open, 45 Days Closed Vial |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
23-285-729
|
Streck
221106 |
1 x 3 mL |
Pack for $281.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
23-285-730
|
Streck
221107 |
2 x 3 mL |
Pack of 2 for $354.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
23-285-732
|
Streck
221108 |
3 x 3 mL |
Pack of 3 for $390.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
Streck Hematology Calibrators are manufactured for calibrating multi-parameter hematology analyzers
- Cal-Chex A Plus Calibrator is a stable suspension of red blood cells, white blood cells and platelets
- Assigned values are derived from replicate analyses on whole-blood calibrated hematology analyzers
- Closed-vial stability: 45 days
- Open-vial stability: 5 days
Specifications
Abbott CELL-DYN™ 1700, 3000, 3200, 3500 and 3700 analyzer | |
Cal-Chex A Plus Hematology Calibrators | |
5 Days Open, 45 Days Closed Vial |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title