Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streamlight™ 3N ProPolymers™ LED Flashlight
SDP

Lightweight model has plenty of power to light your way

Supplier:  Streamlight™ 62202

Catalog No. 19-120-3861



Add to Cart

Description

Description

Features three super-bright, 100,000-hour LEDs. Provides up to 120 hours of run time.

  • Shock-resistant, engineering-grade polymer case
  • Unbreakable polycarbonate lens
  • Push-button switch
  • Nonconductive and corrosion-proof
Specifications
  • Run Time: Up to 120 hours on three “N”alkaline batteries
  • Length: 5.25 in. (13.4cm)
  • Weight 2.1 oz.
  • Color: Yellow

Certifications

UL listed, Class l, Div l, for use in hazardous locations.

Specifications

Specifications

LED Flashlight
100,000 hr.
0.059 kg
Alkaline
Yellow or Black
Polycarbonate
White LED
2.1 oz.
120 hr.
CE and MSHA, UL Class I, II, Div. 1, Groups A,B,C,D,E,F,G; Class III, T4A Classified
Push-button switch
30 lm
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit