Learn More
Dynalon™ Straight-Side PETE Containers
SDPSCI_ED
For storing pastes, samples, powders, specimens etc. in various lab applications. Dynalon™ Straight-Side PETE Containers may be used below freezing to a maximum of 70°C.
Supplier:  Dynalon™ 4263150008
 View more versions of this product
Catalog No. S37263
Item Discontinued This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative.
Description
Description
  • Containers are made of shatterproof, durable PETE with better gas barrier properties with H20, O2 and CO2
  • Transparent with white polypropylene liner less screw closures
Specifications
Specifications
73 mm
8 oz.
PETE
Straight Sided Container
250 mL
Transparent
79.5 mm
Screw Closure
Store Pastes, Samples, Powders, Specimens