Learn More
Eric Armin Incorporated Hanhart Spectron Digital Stopwatch - Calibrated
SDP
Supplier:  Eric Armin Incorporated 2511951VW
Encompass_Preferred
CTRS (Certified Traceable Registration Sticker) - traceable to NIST (National Institute of Standards and Technology). Switchable between 1/100 min and 1/100 sec. 65 memories with fast run memory. Countdown function. Time and date. Split time can be programmed with real time. Water resistant ABS case. Protective pouch with lanyard. 1.5V AAA battery. Battery life: 5-6 years
Catalog No. NC9738819