Learn More
Glas Col LLC Stir Mantle
SDP
Supplier:  Glas Col LLC 100D TEM112M
Encompass_Preferred
3L aluminum housed stir mantle motor drive, u magnet, 100-1850rpm, 500W, 115V.
Catalog No. NC1797852
Due to product restrictions, please to purchase or view availability for this product.