Learn More
Hampton Research Sticky Pad
Supplier: Hampton Research HR4430
Sticky pad is a 7.75 x 6.25 x 0.03 inch sticky pad. Place Sticky Pad on the lab bench, set your crystallization plate on the Sticky Pad and enjoy a non slip crystallization set up and seal experience. Wash to renew grip. As sticky as a Madagascar chameleon tongue, but without the residue, and Sticky Pad is respositionable.