Learn More
Cole-Parmer WBS-300-120 Stuart Shaking Water Bath; 120 VAC, 60Hz
Supplier: Cole-Parmer 1215200
Use this water bath as a linear or orbital shaker, or a static bath by simply changing the platform. Strong magnets couple to the drive unit so platform changes are quick and easy. The stainless steel shaking platforms with high-tension springs accommodate most bottle and flask sizes. It can hold up to 8 x 250 mL, 6 x 500 mL, or 4 x 1 L Erlenmeyer flasks. For tubes and smaller vessels, order an optional stainless steel rack.Set speed and temperature using analog dials. Use the LED display to set and view current temperature. A polycarbonate cover (12152-99) or stainless steel cover is required to reach temperatures above 90°C—order separately. Stainless steel tank with integral drain makes cleaning and maintenance fast and efficient.Please Note: Effective June 2022, the Stuart Shaking Water Bath Model number SBS40/120V/60 has now been rebranded Cole-Parmer under Model number WBS-300-120. All the same great features and specifications, just with a new and improved look.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.