Learn More
Thomas Scientific SteriWare LiquiThief, 1000mm, Sterile, Individually Wrapped
Supplier: Thomas Scientific 1163X01
The SteriWare LiquiThief is simple to use and comes with the added advantage that there is no cleaning validation and no cleaning. The sampler is made in a cleanroom so you simply unwrap and use. If you want to buy a disposable liquid sampler then the LiquiThief is the sampler of choice. The Disposable LiquiThief is a fast and convenient way of taking of liquid samples. It is essentially a big syringe. The operator pulls up the handle to pull the liquid into the sampler. The further the handle is pulled the bigger the sample volume. It is ideal for keg, container and drum sampling. Ideal for sampling low viscosity liquids and oils. Fully welded construction (no adhesives or screws are used) Assembled and packed in a class 100,000 clean room Each sampler is individually wrapped Made from FDA and EU conforming HDPE. All parts are food grade compliant BSE/TSE free Full material certification is available including sterilisation certificates
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.