Learn More
Medicus Health Ergonomic Step Stool
Supplier: Medicus Health 4000M1
∙ Surgery Step Stools - Ergonomic Bright Orange Step Stool brings the work surface to the correct height ∙Ergo-Step Stool is lightweight and easy to clean ∙Stackable step stools bring work surface to the correct height∙Lightweight design makes the step stool easy to move with your feet∙Stools anti-fatigue cushioned standing surface reduces muscle fatigue in feet and legs∙Anti-skid feet keep stool in place while standing on it∙Washable bright orange body provides for high visibility∙Sturdy enough to support up to 500 lbs.∙Constructed of heavy duty plastic ∙Lock the stools together to create a larger standing platform∙Overall Dimensions 18"L x 14"W x 5"H∙Top of the platform dimensions 16-1/2"L x 12-1/2"W∙Cleaning instructions: Clean with standard disinfecting solutions for O.R. equipment or wash in the cart wash with a cycle not to exceed 140°F (60°C)
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.