Learn More
Labconco™ SteamScrubber™ and FlaskScrubber™ Laboratory Glassware Washer Accessory, Utensil Basket
For use with SteamScrubber and FlaskScrubber Laboratory Glassware Washers
Supplier: Labconco™ 4402201/EMD
Description
- Holds small or lightweight labware for safe washing
- Retainer top is required and included
- May be placed in upper or lower Standard Open or Spindle Rack in Labconco FlaskScrubber or SteamScrubber
Warranty and Services
1 year
Specifications
Stainless Steel (304) | |
Washing and drying miscellaneous labware such as stoppers, spatulas and other small lightweight item | |
8 in. | |
4 in. | |
8 in. | |
2 lb. |
Utensil Basket | |
SteamScrubber and FlaskScrubber Laboratory Glassware Washers | |
20.3 cm | |
10.1 cm | |
20.3 cm | |
907 g |