Learn More
XOS Standard XRF Sample Cups - Bag of 100
Supplier: XOS 20033401
Standard XRF Sample Cups and films are the basic necessities for containing and analyzing samples. Used with Sindie, Clora, Signal, Maxine, and Phoebe benchtop analyzers.