Learn More
Cole-Parmer Digi-Sense Panel-Mount Standard Connector, Type-K, Round, Female, 1/Ea
Supplier: Cole-Parmer 1852725
Mount connectors to your panel using the two screws located on the front of the jack. Each connector is ANSI color-coded by thermocouple type: black (J), yellow (K), or blue (T). Choose between flat-bladed miniconnectors and round-pinned standard connectors.