Learn More
ACL Inc Staticide Diamond ESD Paint, 1 Gallon
Supplier: ACL Inc 4700SS 1
Carbon-loaded water-based polyurethane coating for concrete dissipates static discharge for cost-effective static control. Designed for interior concrete floors and unfinished surfaces, this ESD paint is perfectly suited for temporary ESD work spaces, data centers, warehouses, and creating Faraday cages. A truly reliable static dissipative surface, this product conforms to ANSI/ESD S20.20 when grounded and tested according to test method STM 7.1 – 2013. Two-inch-wide copper grounding strips for grounding to earth ground or grounding system are available. Copper is 1.4 mil thickness (2.6 mil thickness with the conductive adhesive backing).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.