Learn More
Stanbio™ Accessories for STAT-Site™ M B-HB Meter (B-Hydroxybutyrate System)
Assurance from false-positives, producing more confidence in results and greater test utility
Supplier: Stanbio™ 303000
Includes: STAT-Site M B-HB Controls, 1 low, 1 high
Description
- Quantitative testing–results in two minutes
- Metabolic status, insulin uptake, and end of catabolism
- STAT differential diagnosis between lacticacidosis and ketoacidosis
- Blood ketone testing for diagnosis and monitoring of ketoacidosis
- Tests for Β-hydroxybutyrate, the most accurate marker for measuring Diabetic Ketoacidosis (DBA)
- Monitors the status of ketoacidosis, documents insulin uptake and defines the end of catabolism
- An alternative to qualitative nitroprusside tests
- Virtually no interference from captopril and other sulfhydryl drugs
- Can also aid in STAT differential diagnosis between lacticacidosis and ketoacidosis
- Use with Stanbio Stat-Site™ Analyzer for convenient, bedside testing
Specifications
STAT-Site M B-HB Controls (strip/card), 1 low, 1 high | |
Blood Ketone testing | |
STAT-Site M B-HB Controls | |
2 x 3 mL |