Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OHAUS™ Starter™ pH Meter ST2200
The Starter™ pH Meter ST2200 is an affordable benchtop pH Meter that clearly displays information with five distinctly marked keys, allowing users to operate with minimal training.
Supplier: OHAUS™ 30656033/EMD
Description
- Equipped with a large 3.5 in. LCD display
- Features a 99 item memory for measurements and last calibration
- Features auto-buffer recognition and auto-endpoint modes that make measurement and calibration simple
- Choose between just the ST2200 meter with electrode holder and a full kit including ST2200 meter, electrode holder, ST320 electrode, 4 pH buffer mini kits (50 mL)
Specifications
±0.01 | |
±0.5°C | |
ST2200 Meter, Electrode Holder | |
Linear | |
0.01 | |
1 to 3 | |
LCD, 3.5 in. Segment Matrix |
±1.5 | |
Datalog for Measurements and Last Calibration | |
Auto-stop, Continuous | |
0 to 14 | |
±1999 | |
0.0°C to 100.0°C | |
99 Sets |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction