Learn More
OHAUS™ Starter™ pH Electrode
The Starter™ pH Electrode features a durable shaft built to withstand daily use. Includes a temperature sensor, which powers automatic temperature compensation and ensures accurate measurements.
Supplier: OHAUS™ 30681115/EMD
Description
- Designed for accurate and precise measurement
- Can be used with both bench and portable meters
- Temperature sensor features automatic temperature compensation
Specifications
BNC | |
Glass | |
5°C to 90°C |
Ceramic | |
2 to 13 | |
pH Electrode |