Learn More
OHAUS™ Starter™ Conductivity Electrode STCON8 with Flow Chamber
The Starter™ Conductivity Electrode STCON8 with Flow Chamber features a glass shaft built to withstand daily use. Includes a temperature sensor, which powers automatic temperature compensation for accurate measurements. Flow chamber produces accurate measurements.
Supplier: OHAUS™ 30681235/EMD
Description
- Designed for accurate and precise measurement
- Can be used with both bench and portable meters
- Flow chamber optimizes the sample flow volume and creates mixing patterns that produce accurate and stable measurements
Specifications
0.1 cm ±0.02 | |
Mini-DIN | |
Glass | |
0.055 to 300 μS/cm |