Learn More
OHAUS™ Starter™ Conductivity Electrode
The Starter™ Conductivity Electrode features a glass shaft built to withstand daily use. Includes a temperature sensor, which powers automatic temperature compensation for accurate measurements.
Supplier: OHAUS™ 30681117/EMD
Description
- Designed for accurate and precise measurement
- Can be used with both bench and portable meters
- Temperature sensor features automatic temperature compensation
Specifications
0.1 cm ±0.02 | |
Mini-DIN | |
0.055 to 300 μS/cm |
Conductivity Electrode | |
Glass |