Learn More
OHAUS™ Starter™ Conductivity Electrode
The Starter™ Conductivity Electrode features a glass shaft built to withstand daily use. Includes a temperature sensor, which powers automatic temperature compensation for accurate measurements.
Supplier: OHAUS™ 30681116/EMD
Description
- Designed for accurate and precise measurement
- Can be used with both bench and portable meters
- Temperature sensor features automatic temperature compensation
Specifications
1 cm ±0.2 | |
Mini-DIN | |
50 to 2000 μS/cm |
Conductivity Electrode | |
Glass |