Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Starplex™ Scientific Starswabs II™ Dry Swabs
$1160.00
Specifications
Shelf Life | 2 Years |
---|---|
Sterility | Sterile |
For Use With (Application) | For assessing microbial presence on various surfaces and equipment |
Product Type | Starswabs II™ Dry Swab |
Catalog Number | Mfr. No. | Packaging | Tip Material | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Packaging | Tip Material | Price | Quantity & Availability | ||||
14-905-50
|
Starplex™ Scientific
S09D |
Tube | Rayon |
Case of 500 for $1,160.00
|
|
||||
Description
Dry, sterile swabs with plastic shafts
Specifications
2 Years | |
Sterile | |
For assessing microbial presence on various surfaces and equipment | |
Starswabs II™ Dry Swab |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title