Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Starplex™ Scientific Starswab II™ Collection and Transport Systems: Amies Clear Medium
Protective overskirt on cap
$108.90 - $1130.00
Specifications
Product Type | Collection and Transport System |
---|---|
For Use With (Application) | Collecting, transporting and maintaining the viability of aerobic, anaerobic and facultative microorganisms |
Packaging | Bag |
Storage Requirements | 4°C to 25°C |
Tip Material | Rayon |
Catalog Number | Mfr. No. | Media Type | Shaft Material | Shelf Life | SwabConfiguration | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Media Type | Shaft Material | Shelf Life | SwabConfiguration | Price | Quantity & Availability | ||
14-905-80
|
Starplex™ Scientific
SP130D |
Modified Amies Clear Gel | Plastic | 2 Years | Dual Applicator |
Pack of 50 for $144.20
Case of 10 PK for $1,162.00
|
|
||
14-905-91
|
Starplex™ Scientific
SP130L |
Liquid Amies | Plastic | 18 Months | Single Applicator |
Case of 500 for $1,028.00
|
|
||
14-905-92
|
Starplex™ Scientific
SP130LD |
Liquid Amies | Plastic | 18 Months | Dual Applicator |
Case of 500 for $1,130.00
|
|
||
14-905-67
|
Starplex™ Scientific
SP130X |
Modified Amies Clear Gel | Plastic | 2 Years | Single Applicator |
Pack of 50 for $108.90
Case of 10 PK for $906.50
|
|
||
14-905-81
|
Starplex™ Scientific
SP130W |
Modified Amies Clear Gel | Straight Wire | 2 Years | Minitip flocked |
Case of 100 for $628.00
|
|
||
Description
- Designed for collecting, transporting and maintaining the viability of aerobic, anaerobic and facultative microorganisms
- Swab tips available in single, double or straight wire format
- Applicators have color-coded caps with overskirt and biohazard plug-cap rings; extra-long swab shafts; and a labeled, round-bottom, spiral transport tube
- Transport tubes contain 4mL of semi-solid media
- Spiral media chamber within a seamless polypropylene tube protects sample and keeps it immersed in media during transport
- Labels preprinted with lot number, expiration date and product code with space reserved for recording patient information
- Gamma sterilized and wrapped in sterile, easy-to-peel envelope
Amies Clear Medium
- General-purpose transport medium for recovery of a wide range of organisms, including certain anaerobes
- Provides excellent survival characteristics for transport and gram stain compatibility
Specifications
Collection and Transport System | |
Bag | |
Rayon |
Collecting, transporting and maintaining the viability of aerobic, anaerobic and facultative microorganisms | |
4°C to 25°C |
Safety and Handling
ShelfLife : 2 Years
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title