Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Starplex™ Scientific Magenta Lined Cap Specimen Container
The original magenta, flat, lined cap is available with a 90ml sterile polypropylene container
Supplier: Starplex™ Scientific B908L
Specifications
Polypropylene | |
90 mL | |
Lined Cap | |
Specimen collection/transportation |
Sterile | |
Magenta | |
White | |
Labeled |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title