Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Starplex™ Scientific Multitrans™ Collection and Transportation Systems

Complete collection and transport systems for viruses, Chlamydia, Mycoplasma and Ureaplasma

$300.50 - $441.00

Specifications

Storage Requirements 2°C to 25°C
View More Specs

Products 2
Catalog Number Mfr. No. Product Type Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Quantity Price Quantity & Availability  
22-046-420
View Documents
Starplex™ Scientific
S1600
Multitrans Collection and Transportation System 100/Cs.
Case of 100 for $441.00
Add to Cart
 
22-046417A
View Documents
Starplex™ Scientific
S160100
Multitrans Collection and Transportation System 50 Pk.
Pack of 50 for $300.50
Case of 2 PK for $509.50
Add to Cart
 
Description

Description

Complete collection and transport systems for viruses, Chlamydia, Mycoplasma and Ureaplasma

  • Consist of a scored plastic shafted Dacron™-tipped swab, transport medium with glass beads and antibiotics, and a 10mL clarified polypropylene leak-resistant tube with skirted screw cap
  • Color-coded cap prevents mix-ups

Specifications

Specifications

2°C to 25°C
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit