Learn More
Boekel Scientific™ Standard Tissue Flotation Bath PROMO
Prevents wrinkles and distortion during preparation of paraffin embedded tissue sections.
Manufacturer:  Boekel Scientific™ 14792PM
Catalog No. 15-464-30PM
Add to cart
This item is not returnable. View return policy
Description
Description
  • Edges serve as convenient drying surface for processed slides
  • Bath surface is black ceramic coated steel for improved visibility of tissue sections
  • Supplied with dust cover, thermometer holder and non-skid rubber feet
  • Equipped with on/off switch, pilot light and thermostat
  • Durable and inexpensive
  • Rapid heat-up
  • Easy maintenance
Warranty and Services
Two Years
Specifications
Specifications
35°C to 65°C
3.75 in.
11 in.
5 lb.
115 V
50/60 Hz
Flotation Bath
9.2 cm
27.9 cm
2.3 kg
190 W