Learn More
Bal Supply Standard Taper Joint Clips, Keck Type, POM
Will not scratch glass. Color coded for easy size identification.
Supplier: Bal Supply 203112
Description
- Made from Polyoxymethylene (POM)
- Will not scratch glass
- Color-coded
- For standard taper joints
Specifications
Autoclavable | |
Standard Taper Joint Clip | |
12 | |
Standard Taper |
10 Pk. | |
POM | |
Violet | |
Non-sterile |