Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Public Seating™ Standard Steel Folding Chairs
SCI_ED

Seat has waterfall front for greater comfort.

Supplier:  National Public Seating™ 051

Catalog No. S98038MF

Item Discontinued This item has been discontinued by Fisher Scientific. Please to view product availability in your area.

This item is not returnable. View return policy

Description

Description

  • 19-Gauge, 0.875 in. (2.2cm) round tubular frame
  • 2.5 in. (6.4cm) frame strengtheners on each side of seat
  • Two 19-gauge, 0.625 in. (1.6cm), U-shaped, double-riveted cross braces
  • Back features single contour with curled safety edges
  • Back and frame electromagnetically welded together
  • Color-coordinated V-Tip plus for added stability
  • Dimensions (folded): 2D × 18.75W × 38 in.H (5 × 48 × 97cm)

Order Info

  • Ships four chairs per carton; 40 lb. (18.1kg)

Specifications

Specifications

Folding Chair
18.75 in.
40 cm
15.75 in.
74.9 cm
52.7 cm
Chair
Beige
40.64 cm
47.6 cm
16 in.
20.7 in.
29.5 in.
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit