Learn More
American Biotech Supply Standard Hazardous Location (Explosion Proof) Freezer with Natural Refrigerants
Freezers are engineered for cold-storage use in environments where volatile conditions could potentially exist. These units meet NFPA and OSHA guidelines Class 1, Division II, Group C & D for storing substances in hazardous locations.
$5591.00 - $6778.00
Specifications
Type | HazLoc Freezer |
---|---|
Temperature Range | -15°C to -25°C |
Temperature Sensor | Microprocessor Temperature Controller |
Temperature Stability | ±0.1°C |
Door Style | Solid Swing |
Catalog Number | Mfr. No. | Capacity (English) | Capacity (Metric) | Amperage | Min. Clearance | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (English) | Capacity (Metric) | Amperage | Min. Clearance | Price | Quantity & Availability | ||
22-651-386
|
American Biotech Supply
ABTHCEFP14 |
14 cu. ft. | 396 L | 3.5 A | 75 in. |
Each for $5,591.00
|
|
||
22-651-387
|
American Biotech Supply
ABTHCEFP17 |
17 cu. ft. | 481 L | 6 A | 70 in. |
Each for $6,234.00
|
|
||
22-651-388
|
American Biotech Supply
ABTHCEFP20 |
20 cu. ft. | 566 L | 6 A | 80 in. |
Each for $6,778.00
|
|
||
Description
Standard Hazardous Location (Explosion Proof) Freezers with Natural Refrigerants are engineered for cold-storage use in environments where volatile conditions could potentially exist. All units include a two year parts and labor warranty. They run on natural, hydrocarbon refrigerant for environmental health and economic efficiency. These units meet NFPA and OSHA guidelines Class 1, Division II, Group C & D for storing substances in hazardous locations.*
*Explosion Proof Units are designed for environments where external hazards could exists. For storage of volatile substances in normal laboratory conditions, see ABS's Flammable Storage Units.
- Microprocessor Temperature Controller
- Interior shelving
- Keyed door lock
- High density urethane foam cabinet and door insulation
- Not equipped with line cord and plug; must be hardwired in conduit to 120 VAC power source
- ETL Listed
Specifications
HazLoc Freezer | |
Microprocessor Temperature Controller | |
Solid Swing | |
Right | |
Polyurethane Foam | |
Powder-coated Steel | |
cULus | |
Manual | |
Fan-forced | |
None | |
Manual | |
No Plug, Hardwired | |
Bottom-mounted |
-15°C to -25°C | |
±0.1°C | |
1 | |
Scratch-resistant Painted Steel | |
Plastic | |
6 Fixed | |
Upright | |
White | |
Hazardous Location Cold Storage | |
Two Year Parts and Labor Warranty | |
Microprocessor Temperature Controller | |
115 V | |
Natural, Hydrocarbon |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.