Learn More
Custom Biogenic Systems STANDARD DIVIDER, 49-CELL
Supplier: Custom Biogenic Systems D49
7 x 7 matrix divider for holding up to 49 vials or tubes in a freezer box used with an Eppendorf Innova Ultra Low Temperature Freezer (sold separately) • Accommodates a maximum tube diameter of 17.4cm (0.69")