Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stanbio™ Sirrus™ Analyzer Standards and Calibrators

$116.60 - $334.00

Specifications

Content And Storage 2°C to 8°C
View More Specs

Products 2
Catalog Number Mfr. No. Type Quantity For Use With (Equipment) Stability Price Quantity & Availability  
Catalog Number Mfr. No. Type Quantity For Use With (Equipment) Stability Price Quantity & Availability  
SB0595003
View Documents
Stanbio™
0595003
Direct HDL/LDL Cholesterol Calibrator 1 x 3 mL Sirrus clinical chemistry analyzer 21 Days
Pack for $116.60
Add to Cart
 
SB0365401
View Documents
Stanbio™
0365401
Direct Glycohemoglobin Calibrator Set 4 x 0.5 mL Stanbio sirrus clinical analyzer 1 Month
Pack for $334.00
Add to Cart
 
Description

Description

  • For use with Stanbio Sirrus Clinical Analyzers, Cat. Nos. SBG-550-0001, SBG-500-0001

Specifications

Specifications

2°C to 8°C
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit