Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Stanbio™ LiquiColor™ Enzymatic Cholesterol Test
For total and HDL cholesterol assays
Supplier: Stanbio™ 1010225
Includes: Standard: 1 × 3mL, reagent: 2 × 250mL
Description
- Ready-to-use liquid reagent
- With lipid clearing factor
Specifications
Cholesterol LiquiColor™ Test | |
Total and HDL Cholesterol assays | |
Standard: 1 x 3mL, reagent: 2 x 250mL, lipid clearing factor | |
2 x 250 mL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title